missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ West Nile Virus MP Antibody Blocking Peptide

Catalog No. NB398661
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398661 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398661 Supplier Novus Biologicals™ Supplier No. NB10056743PEP
Only null left

Specifications

Concentration 1 mg/mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NB100-56743
Quantity 0.1 mg
Research Category Immunology, Virology Bacteria and Parasites
Storage Buffer Peptide dissolved in dH20. Contains no BSA.