missing translation for 'onlineSavingsMsg'
Learn More

New Pig Wesco™ Folding Hand Truck

Hand-cart your boxes and other loads while saving storage space with this folding hand truck.

Supplier:  New Pig MTL713

 View more versions of this product

Catalog No. 17200128


Only null left

Description

Description

  • Loading capacity up to 550 lb.
  • Rubber Tread Poly Hub.
  • Noseplate
Specifications

Chemical Identifiers

Fold-up Hand Truck
Dolly
Aluminum

Specifications

Fold-up Hand Truck
550 lb.
Aluminum
23.5 x 18 in.
Dolly
249.5 kg
Black
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions