missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ Thermofill™ Lined Leather Palm Gloves

Catalog No. 19148751
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19148751 X-Large
19148749 Medium
19148750 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19148751 Supplier Wells Lamont™ Supplier No. Y0042XL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pigskin Leather Palm Gloves provide comfort and thermal protection with Thermofill™ lining. The knuckle strap and safety cuff provide additional protection for the knuckles and wrist.

  • Premium pigskin leather palm
  • Thermofill™ lined
  • Safety cuff
  • Wing thumb
  • Gunn cut

Specifications

Cuff Material Fabric
Color Tan
Grade Premium
Hand Specific
Material Pigskin Leather
Product Type Thermal Lined Gloves
Size X-Large
Style Gunn
For Use With (Application) Ideal in the outdoors, freezers or any cold environments.
Glove Cuff Color Blue with Yellow Stripes
Glove Cuff Style Safety
Hem Color Blue
Liner Material Thermofill™
Series Y0042
Thumb Style Wing
Show More Show Less