missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ Premium Grain Cowhide Leather Palm Work Gloves

Catalog No. 1146232C
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
1146232C Large
1146232D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1146232C Supplier Wells Lamont™ Supplier No. Y2009L
Only null left

Premium Grain Cowhide Leather Palm Work Gloves are constructed of cow grain leather with a full leather index finger, leather tips, and a leather knuckle strap. The lined palm provides additional comfort in the palm of the hand.

  • Premium quality grain cowhide
  • Grain knuckle strap
  • Heavy duty canvas back
  • Gauntlet cuff
  • Wing thumb
  • Gunn cut

Specifications

Back Material Canvas
Color Natural, Blue
Grade Premium
Hand Specific
Material Grain Cowhide Leather
Product Type General Purpose Gloves
Size Large
Style Gunn
Glove Cuff Color Blue
Glove Cuff Style Gauntlet
Hem Color Brown
Palm Type Lined
Series Y2009
Thumb Style Wing
Show More Show Less