missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ Standard Shoulder Split Leather Work Gloves

Standard Shoulder Split Leather Work Gloves are durable gloves offering split leather index fingers, leather tips, and a leather knuckle strap with a lined palm for additional comfort.

$61.40 - $61.40

Specifications

Color Gray, Red
Grade Standard
Hand Specific
Material Shoulder Split Cowhide Leather
Style Gunn
View More Specs

Products 1
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19148814
View Documents
Wells Lamont™
Y3201M
Medium
Pack of 12 for $61.40
Case of 6 Pack for $352.14
Only null left
 
Description

Description

  • Standard shoulder split cowhide leather palm
  • Full leather palm, fingertips and knuckle strap
  • Striped canvas back and cuff
  • Wing thumb
  • Safety cuff
  • Lined palm
Specifications

Specifications

Gray, Red
Specific
Gunn
Safety
Standard
Shoulder Split Cowhide Leather
Everyday work glove recommended for the Warehousing and Construction industries.
Wing
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'