missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ Spec-Tec™ Sterile Cut Resistant Glove Liners - ANSI A1

Catalog No. 19048680
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19048680 Small
19048681 Medium
19048682 Large
19048685 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19048680 Supplier Wells Lamont™ Supplier No. M102S
Only null left

HPPE fibers are comfortable and stretchy, while also providing ANSI Cut Level A1 protection. Each pair of Spec-Tec™ Sterile Cut Resistant Glove Liners is individually wrapped in a sterile package.

Cut resistant liners with tactile sensitivity

  • 100% HPPE fiber construction
  • Liners provide ANSI Cut Level A1 protection
  • Individually wrapped in sterile packaging
  • Made in the USA

Specifications

ANSI Abrasion Rating ANSI 1
Coating Material Uncoated
Color White
Cut Resistance Rating ANSI A1
Disposable Reusable
Hand Ambidextrous
Latex Free Yes
Material HPPE
Product Type Cut-resistant Glove Liners
Size Small
Sterility Sterile
Finger Style Full
For Use With (Application) Medical, Pharmaceuticals, and Organ Tissue Processing
Glove Cuff Style Knit
Hem Color Green
Inspected for Medical Use Yes
Packaging Type Individually wrapped in a sterile package.
Series M102
Show More Show Less