missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ Missouri Mule Premium Select Split Cowhide Leather Welder's Gloves

Catalog No. 1146245C
Change view
Click to view available options
Size:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
1146245C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1146245C Supplier Wells Lamont™ Supplier No. Y1903L

missing translation for 'validMainframeMsg'

Missouri Mule Premium Select Split Cowhide Leather Welder's Gloves are sewn with Aramid Fiber for maximum protection against hazards such as heat, flames, and sparks.

  • Select side split premium cowhide leather construction
  • Sewn with Aramid Fiber thread for extreme strength and wearability
  • Fully welted to add durability, and to reduce the risk of burning through the seams
  • Fleece jersey lining for enhanced comfort and heat protection
  • Gunn cut, reinforced thumb

Specifications

Color Brown
Cut Resistance Rating ANSI A2
Grade Premium
Material Side Split Cowhide Leather
Product Type Welding Gloves
Size Large
Style Gunn
For Use With (Application) Construction, Steel/Metal Work, and Welding
Glove Cuff Style Gauntlet
Liner Material Fleece
Puncture Rating ANSI/ISEA 105-16 Level 3
Series Y1903
Thumb Style Reinforced
Show More Show Less