missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ Full-Finger Medical Nylon Glove Liners - Extended Cuff

Catalog No. p-136845
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19044153 Small
19044154 Medium
19044155 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19044153 Supplier Wells Lamont™ Supplier No. M113 S
Only null left

Liners are constructed of high-quality, 100% continuous filament nylon and packaged in a secure box. Wear under a disposable glove for additional comfort, or to reduce hand fatigue from perspiration.

  • 100% continuous nylon filament creates an ideal barrier between hands and disposable gloves
  • Reduces allergies and adverse reactions that can be caused by prolonged exposure to latex
  • Packaged in a secure box so that liners remain clean
  • Fully launderable up to 20 times without shrinking
  • Full finger, extended cuff design
  • Non-sterile
  • Made in the USA

Specifications

Applications Healthcare
Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Latex Free Yes
Material Nylon
Product Type Nylon Glove Liner
Size Small
Sterility Non-sterile
Finger Style Full
For Use With (Application) Reducing Latex Allerigies and Adverse Reactions
Glove Cuff Style Knit
Hem Color Green
Packaging Type Box
Series M113
Show More Show Less