missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ MechPro™ Synthetic Leather Palm Mechanics Gloves

Catalog No. 191305873
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
191305873 Small
191305874 Medium
19099240C Large
191305876 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 191305873 Supplier Wells Lamont™ Supplier No. 7701S

MechPro™ Synthetic Leather Palm Mechanics Gloves are constructed of heavy duty, ribbed knit fabric for abrasion and incidental heat resistance. Double stitched in high wear areas, with a gusseted finger crotch and perforated finger fourchettes for comfort.

  • Gloves provide grip and dexterity for handling tools or parts
  • Ribbed knit fabric for nick, abrasion, and incidental heat resistance
  • Synthetic leather palm and fingertips
  • Reinforced fingertips with pinched fingertip design
  • Gusseted finger crotch and perforated finger fourchettes for comfort
  • Double stitched in high wear areas: fingertips, index finger, and palm
  • Elastic cuff with Feeltite™ hook and loop adjustable wrist closure

Specifications

Back Material Ribbed Knit
Construction Knit
Cuff Material Elastic
Coating Material Uncoated
Color Black
Hand Specific
Material Synthetic Leather
Product Type Mechanic's Gloves
Size Small
For Use With (Application) Fleet/Maintenance Mechanics, Pipe Fitters, Electricians, Millwrights, and Various Trades Industries
Glove Cuff Style Hook and Loop
Series 7701
Show More Show Less