missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ Economy Grade Grain Cowhide Leather Drivers Gloves

Economy Grade Grain Cowhide Leather Drivers Gloves with a gunn cut and straight thumb design to offer comfort and durability.

$59.12 - $67.11

Specifications

Color Natural
Grade Economy
Hand Specific
Material Grain Cowhide Leather
Product Type Driver Gloves
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19044200
View Documents
Wells Lamont™
Y0135S
Small
Dozen of 12 for $59.12
Case of 12 Dozen for $650.40
Only null left
 
19044201
View Documents
Wells Lamont™
Y0135M
Medium
Dozen of 12 for $59.12
Case of 12 Dozen for $650.40
Only null left
 
19044202
View Documents
Wells Lamont™
Y0135L
Large
Dozen of 12 for $59.12
Case of 12 Dozen for $650.40
Only null left
 
19044203
View Documents
Wells Lamont™
Y0135XL
X-Large
Dozen of 12 for $67.11
Case of 12 Dozen for $738.12
Only null left
 
Description

Description

  • Economy choice
  • Gunn cut
  • Straight thumb
  • Bound hem
Specifications

Specifications

Natural
Specific
Driver Gloves
Open
Straight
Economy
Grain Cowhide Leather
Gunn
Y0135
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'