missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ GuardTec3™ Hi-Vis Foam Latex Palm Gloves

$127.52 - $127.52

Specifications

Color Gray, Orange
Cut Resistance Rating ANSI A3
Material Nitrile, FlexTech
Product Type Cut-resistant Gloves
For Use With (Application) Cut protection, high visibility applications
View More Specs

Products 6
Catalog Number Mfr. No. Glove Cuff Color Size Price Quantity  
Catalog Number Mfr. No. Glove Cuff Color Size Price Quantity  
17200305
View Documents
Wells Lamont™
Y9282HVXS
Yellow X-Small
Dozen of 12 Each for $127.52
Only null left
 
17200306
View Documents
Wells Lamont™
Y9282HVS
Red Small
Dozen of 12 Each for $127.52
Only null left
 
17200307
View Documents
Wells Lamont™
Y9282HVM
Green Medium
Dozen of 12 Each for $127.52
Only null left
 
17200308
View Documents
Wells Lamont™
Y9282HVL
Brown Large
Dozen of 12 Each for $127.52
Only null left
 
17200309
View Documents
Wells Lamont™
Y9282HVXL
Blue X-Large
Dozen of 12 Each for $127.52
Only null left
 
17200310
View Documents
Wells Lamont™
Y9282HVXXL
Black 2X-Large
Dozen of 12 Each for $127.52
Only null left
 
Description

Description

Specifications

Specifications

Gray, Orange
Nitrile, FlexTech
Cut protection, high visibility applications
ANSI A3
Cut-resistant Gloves
Knit
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'