missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ Grain Goatskin Leather Drivers Gloves

Catalog No. 19821953
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19821953 X-Large
19821950 Small
19821951 Medium
19821952 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19821953 Supplier Wells Lamont™ Supplier No. Y0107XL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Grain Goatskin Leather Drivers Gloves feature goatskin leather, the thinnest material available to provide comfort and protection while on the job. Ideal for use in industries such as warehousing, construction, and oil and gas.

  • Quality goatskin with natural lanolin for softness and wear
  • Goatskin has a higher natural tensile strength despite abrasion
  • Gunn cut and keystone thumb for added dexterity and flexibility

Specifications

Color Natural
Hand Specific
Material Grain Goatskin Leather
Product Type Driver Gloves
Size X-Large
Style Gunn
For Use With (Application) Ideal for use in industries such as warehousing, construction, and oil and gas.
Glove Cuff Color Blue
Glove Cuff Style Open
Hem Color Blue
Series Y0107
Thumb Style Keystone
Show More Show Less