Learn More
Wells Lamont™ Grain Goatskin Leather Drivers Gloves
Grain Goatskin Leather Drivers Gloves feature goatskin leather, the thinnest material available to provide comfort and protection while on the job. Ideal for use in industries such as warehousing, construction, and oil and gas.
Supplier: Wells Lamont™ Y0107L
Description
- Quality goatskin with natural lanolin for softness and wear
- Goatskin has a higher natural tensile strength despite abrasion
- Gunn cut and keystone thumb for added dexterity and flexibility
Specifications
| Natural | |
| Grain Goatskin Leather | |
| Large | |
| Ideal for use in industries such as warehousing, construction, and oil and gas. | |
| Brown | |
| Keystone |
| Specific | |
| Driver Gloves | |
| Gunn | |
| Open | |
| Y0107 |