missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ Premium Grain Cowhide Leather Palm Gloves

Catalog No. 1146241D
Change view
Click to view available options
Size:
X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
1146241D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1146241D Supplier Wells Lamont™ Supplier No. Y2008XL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Premium Grain Cowhide Leather Palm Gloves are constructed with cow grain leather on the index finger, finger tips, and knuckle strap. The lined palm provides additional comfort in the palm of the hand.

  • Premium quality grain cowhide leather
  • Grain knuckle strap
  • Heavy duty canvas back
  • Gunn cut
  • Safety cuff
  • Wing thumb

Specifications

Cuff Material Canvas
Color Natural
Grade Premium
Hand Specific
Material Grain Cowhide Leather
Product Type Leather Palm Gloves
Size X-Large
Style Gunn
Glove Cuff Color Blue
Glove Cuff Style Safety
Hem Color Blue
Thumb Style Wing
Show More Show Less