missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ FlexTech™ Y9275 Polyurethane-Coated HPPE Cut-Resistant Gloves

Catalog No. 17200316
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17200316 2X-Large
17200311 X-Small
17200312 Small
17200313 Medium
17200314 Large
17200315 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17200316 Supplier Wells Lamont™ Supplier No. Y9275XXL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

FlexTech™ Y9275 Polyurethane-Coated HPPE Cut-Resistant Gloves with High Performance Polyethylene (HPPE) fibers provide cut protection, while polyurethane coating on the palm/fingers enhances grip and tactility.

  • FlexTech high performance polyethylene fibers (HPPE) and nylon blended shell
  • Gloves provide a balance of comfort, dexterity and cut resistance
  • Polyurethane palm coating enhances grip and tactility
  • Ideal for general assembly and other applications where cut resistance is required
  • Launderable for a longer wear life

Specifications

ANSI Abrasion Rating ANSI 6
Certifications/Compliance ASTM F1790-97, ASTM F2992-15, ASTM D3389-10
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free Yes
Material HPPE
Product Type Cut-resistant Gloves
Size 2X-Large
EN388 Standard EN 388:4342
For Use With (Application) Auto assembly, engine assembly, construction, metal fabrication
Glove Cuff Style Knit
Model Y9275XXL
Puncture Rating ANSI/ISEA 105-16 Level 3
Series FlexTech™ Y9275
Yarn Gauge 13
Show More Show Less

WARNING: Cancer – www.P65Warnings.ca.gov