missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ FlexTech™ Y9287 Polyurethane Coated Nylon Work Gloves

Catalog No. 19148865
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19148865 3X-Large
19148845 X-Small
19148861 Small
19148862 Medium
19148863 Large
19148864 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19148865 Supplier Wells Lamont™ Supplier No. Y9287XXL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

FlexTech™ Y9287 Polyurethane Coated Nylon Work Gloves are ideal for a variety of applications that require cut protection, along with high levels of dexterity. The glove's non-slip surface allows workers to work efficiently with confidence.

  • Synthetic seamless knit shell is comfortable and breathable
  • Polyurethane palm coating offers tactility and enhances grip
  • Provides additional protection to high wear/high risk areas of the hand
  • Dark coloring hides dirt and grime to extend the glove's useful life

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance ANSI/ISEA 105-2024 Cut Level A1, ANSI/ISEA 105-2024 Abrasion Level 4, ANSI/ISEA 105-2024 Puncture Level 2, EN 388:2016+A1:2018 3131
Certifications/Compliance (Europe) EN 388:2016+A1:2018 3131
Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size 3X-Large
For Use With (Application) Electronic Assembly, and Small Parts Handling
Gender Unisex
Glove Cuff Style Knit
Grip Design Smooth
Series Y9287
Yarn Gauge 13
Product Line FlexTech
Show More Show Less