missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ Grain Cowhide Leather Drivers Gloves, Gunn Cut

Grain Cowhide Leather Drivers Gloves featuring Gunn cut and straight thumb design offer comfort with durability and extended wear. Ideal for use in industries such as construction, farming, and warehousing.

$133.79 - $137.91

Specifications

Color Natural
Grade Economy
Hand Specific
Material Grain Cowhide Leather
Product Type Driver Gloves
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19050021A
View Documents
Wells Lamont™
Y0133S
Small
Dozen of 12 Pair for $133.79
Only null left
 
19050021B
View Documents
Wells Lamont™
Y0133M
Medium
Dozen of 12 Pair for $133.79
Only null left
 
19050021C
View Documents
Wells Lamont™
Y0133L
Large
Dozen of 12 Pair for $133.79
Only null left
 
19050021D
View Documents
Wells Lamont™
Y0133XL
X-Large
Dozen of 12 Pair for $137.91
Only null left
 
Description

Description

  • Economy choice
  • Bound hem
  • Straight thumb
  • Gunn cut
Specifications

Specifications

Natural
Specific
Driver Gloves
Open
Straight
Economy
Grain Cowhide Leather
Gunn
Y0133
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'