Learn More
Thomson Instrument Company Well Plate Lid | Use w/ 96 & 24 Well Plates | CS100
Catalog No. 502221372
Encompass
Thomson Instrument Company Well Plate lid fits the entire offering of 24 and 96-well configurations with V, pyramid, or U bottom, and square or round wells for cell culture and analytical sample preparation. Lid is clearand designed to work with SLAS specifications. Many Thomson well plates are individually wrapped for your convince, and ideally designed to work with Thomson filter plates. Thomson manufactures its products in the USA to assure same day shipping.
Catalog No. 502221372 Supplier Thomson Instrument Company Supplier No. 981945
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Thomson Instrument Company Well Plate lid fits the entire offering of 24 and 96-well configurations with V, pyramid, or U bottom, and square or round wells for cell culture and analytical sample preparation. Lid is clearand designed to work with SLAS specifications. Many Thomson well plates are individually wrapped for your convince, and ideally designed to work with Thomson filter plates. Thomson manufactures its products in the USA to assure same day shipping.