Learn More
Thermo Scientific™ Well Plate Accessories
Effectively seal all microplate formats to minimize evaporation and protect samples from contamination and spilling, ensure a consistent, reliable seal with all microplate formats with this easy-to-use, handheld applicator, and get significantly quicker dry-down times than standard methods, and remove most chromatography solvents in just minutes with easy to use 96-well sample concentrators.
$62.60 - $664.18
Specifications
No. of Wells 96
View More Specs
Products 10
Catalog Number Mfr. No. Product Type Volume (Metric) Working Well Shape Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Volume (Metric) Working Well Shape Price Quantity & Availability  
60105424
View Documents Promotion Details
Thermo Scientific™
60180 V109
Deep Well Plate Vial 350 μL Taper
Pack of 960 Each for $664.18
Only null left
60105400
View Documents Promotion Details
Thermo Scientific™
60180V100
Microtiter Plate Vial 500 μL Taper
Pack of 500 Each for $482.17
Only null left
60105401
View Documents Promotion Details
Thermo Scientific™
60180V101
Microtiter Plate Vial 700 μL Taper
Pack of 500 Each for $280.10
Only null left
60105425
View Documents Promotion Details
Thermo Scientific™
60180V110
Square Well Plate Vial 1000 μL Round
Pack of 960 Each for $501.58
Only null left
60105403
View Documents Promotion Details
Thermo Scientific™
60180V104
Microtiter Plate Vial 1100 μL Round
Pack of 500 Each for $409.48
Only null left
60105405
View Documents Promotion Details
Thermo Scientific™
60180V106
Deep Well Plate Vial 1500 μL Round
Pack of 500 Each for $528.75
Only null left
03252446
View Documents Promotion Details
Thermo Scientific™
60180M999
Mat Applicator Not Applicable Not Applicable
Pack for $68.72
Only null left
03252445
View Documents Promotion Details
Thermo Scientific™
60180M950
Sealing Tape Applicator Not Applicable Not Applicable
Pack of 2 Each for $183.75
Only null left
60105432
View Documents Promotion Details
Thermo Scientific™
60180 P407
Tube Tray Not Applicable Not Applicable
Pack of 10 Each for $347.09
Only null left
60105427
View Documents Promotion Details
Thermo Scientific™
60180C100
Center Hole Cap Not Applicable Not Applicable
Pack of 96 Each for $62.60
Only null left
Description
Description
Effectively seal all microplate formats to minimize evaporation and protect samples from contamination and spilling, ensure a consistent, reliable seal with all microplate formats with this easy-to-use, handheld applicator, and get significantly quicker dry-down times than standard methods, and remove most chromatography solvents in just minutes with easy to use 96-well sample concentrators.
Sealing Tape Applicator
Ensure a consistent, reliable seal with all microplate formats and all types of tape with this easy to use, handheld applicator for Thermo Scientific™ WebSeal Sealing Tapes. They are packaged two per box.
Sample Concentrators
Remove most chromatography solvents in just minutes. Simple to operate and maintain, Thermo Scientific™ WebSeal Sample Concentrators allow quick, easy solvent evaporation from 96 well microplates, offering significantly quicker dry-down times than standard methods such as vacuum ovens or centrifuges. These evaporators offer significant throughput advantages for laboratories looking to accelerate microplate sample preparation. Depending on your throughput requirements, choose either the manually operated MiniVap Concentrator or the programmable, robot-ready UltraVap.
Specifications
Specifications
96
Videos
SDS
Documents
Documents
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.