Research Products Corporation Well Orienter Cards, 4 per Package
Catalog No. 50476563
The Well Orienter System increases accuracy and efficiency with 96 well micro-plate procedures. The plastic well index card fits perfectly under a micro-plate and clearly identifies each well with distinct alpha/numeric characters.
Catalog No.
50476563
Supplier
Research Products CorporationSupplier No.
248804
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Well Orienter System increases accuracy and efficiency with 96 well micro-plate procedures. The plastic well index card fits perfectly under a micro-plate and clearly identifies each well with distinct alpha/numeric characters.