missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Welch Vacuum Pump

Catalog No. 16985654
Change view
Click to view available options
Voltage:
220/240 V
115 V
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
16985654 115 V
16985653 220/240 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16985654 Supplier Labconco™ Supplier No. 7584500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemically resistant vacuum pump is used for lyophilization or concentration of samples containing harsh chemicals like TFA, Acetonitrile, etc.

  • This chemically resistant vacuum pump is used for lyophilization or concentration of samples containing harsh chemicals like TFA, Acetonitrile, etc.
  • An alumina filter element neutralizes acids and removes reaction products.
  • Internal oil case is PTFE coated.

Specifications

Flow Rate 158 L/min
Pump Type Rotary Vane Vacuum Pump
Enclosure Rating IP54
Maximum Pressure 0.002 mBar
Voltage 115 V