Learn More
Welch™ Oil-Mist Eliminator for Vacuum Pumps
Accessories for vacuum pumps
Supplier:  Welch™ 1416D
Catalog No. 01-108A
Only null left
Explore available promotions
Description
Description
Oil-Mist Eliminator
  • Hermetically sealed
  • SS w/borosilicate microfiber element, flouroelastomer gaskets
  • Prevents corrosive exhaust gases from escaping
  • Accepts 13/16 in. hose
  • 12.7W x 19.68 in. H
Pump-Adapter Kit
  • For use with Oil-Mist Eliminator
Specifications
Specifications
Prevents corrosive exhaust gases from escaping
19.68 in.
12.7 in.
Oil-Mist Eliminator
1400N, 1402N, 1376N
49.98 cm
13/16 in.
Product Suggestions
Product Suggestions