Learn More
Welch™ DuoSeal™ Pump Oil
Catalog No. p-3070925
Change view
Click to view available options
Volume (Metric):
1 L
3.78 L
18.92 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
01104A 1 L
01104B 3.78 L
01104C 18.92 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01104A Supplier Welch™ Supplier No. 1407K11
Only null left
For use in DuoSeal High-Vacuum Pumps
For use in DuoSeal High-Vacuum Pumps. One gallon.Specially fractionate oil has properties enabling highest performance for DuoSeal pumps
  • Low vapor Pressure
  • High Viscosity
Specifications
Product Type Vaccum Pump Oil
For Use With (Application) Provides the lubrication that helps enable renowned DuoSeal™ pump durability and ensure the highest vacuum performance
For Use With (Equipment) DuoSeal™ Belt-driven pumps
Quantity 1 Ea.
Volume (English) 0.26 gal.
Volume (Metric) 1 L