Learn More
Welch™ Vacuum Pump On/Off Foot Switch
Vacuum pump accessory that provides hands-free operation of Welch DryFast diaphragm and WOB-L piston pumps.
Supplier:  Welch™ 1430C
 View more versions of this product
Catalog No. 0105517
Only null left
Description
Description
  • Hands-free on/off pump operation
  • No adaptors or special connection needed
  • Plugs directly into the power source outlet
  • Switch logic and power varies depending on model:
    • Parts 01-104-17 and 01-055-16: 3-prong U.S. standard plug with 8 ft. cord (CSA, NEMA, and UL Enclosure Type 1)
    • Part 01-055-17: Male and female IEC connections with 8 ft. cord (plugs directly into the IEC connection on the pump)
Specifications
Chemical Identifiers
Foot Switch
Specifications
90 to 230 V
WOB-L piston pumps
Momentary On/Off switch logic
Foot Switch
Product Suggestions
Product Suggestions