missing translation for 'onlineSavingsMsg'
Learn More

Wearwell™ Worksafe™ Anti-fatigue Floor Mat

High performance, versatile drainage mats for use in wet and dry work areas

Supplier:  Wearwell™ 477.78X3X4GRBK

 View more versions of this product

Catalog No. 18999847


Only null left

Description

Description

  • With optional 2 in. wide safety bevel unattached to mats in colors orange, yellow or black
  • Available as single workstation or can be attached together with connector strips to form any size
  • Grease resistant natural rubber
  • Dry or wet conditions
  • Versatile configuration
Specifications

Chemical Identifiers

WorkSafe Grease-resistant
Anti-fatigue Floor Mat
2.2 cm
Black
0.87 in.

Specifications

3 x 4 ft.
2.2 cm
WorkSafe Grease-resistant
0.87 in.
Anti-fatigue Floor Mat
Black
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions