missing translation for 'onlineSavingsMsg'
Learn More

Wearwell™ WorkRite™ Mats

Catalog No. 191303543
Change view
Click to view available options
Dimensions (L x W):
3 x 5 ft.
3 x 10 ft.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
191303543 3 x 10 ft.
191303542 3 x 5 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 191303543 Supplier Wearwell™ Supplier No. 478.123X10GY

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Dimensions (L x W) 3 x 10 ft.
Material CFR Rubber
Color Gray
For Use With (Application) Wet, dry or greasy areas