Learn More
Wearwell™ UltraSoft Diamond-Plate Mat
Antifatigue mat designed for ultimate comfort and performance
Supplier:  Wearwell™ 414.1516X3X10BK
 View more versions of this product
Catalog No. 191500081
Only null left
Description
Description
Antifatigue mat designed for ultimate comfort and performance
  • For heavy use areas, mats resist abrasion and most chemical spills
  • Double thick sponge base made from 0.94 in. (2.4cm) immediate recovery Nitricell™ sponge
  • Specially formulated blend of nitrile rubber and PVC
  • Slip resistant, diamond embossed vinyl surface resists chemicals, abrasion and fire
  • All edges are beveled for safety and convenience
  • Recycled: Item contains recycled content as described in the item detail
Order Info
Optional colored borders available for maximum safety awareness. Contact your Fisher Scientific Customer Service Representative.
Warranty
Three year conditional.
Specifications
Chemical Identifiers
Double-thick Nitricell Sponge
Specifications
10 x 3 ft.
Double-thick Nitricell Sponge
Black
Product Suggestions
Product Suggestions