Learn More
Wearwell™ Tuf Sponge Mat
For use in all dry work areas
Supplier:  Wearwell™ 451.38X3X5GY
 View more versions of this product
Catalog No. 19042712
Only null left
Description
Description
  • 0.38 in. (0.95cm) blown vinyl sponge
  • Ergonomically designed to reduce floor hardness up to 50%
  • Easily cut to fit work areas
  • Edges are rounded for safety
Order Info
Available in cut lengths to 2 ft., 3 ft. and 4 ft. up to 60 ft. (18.3cm). Contact your Fisher Scientific Customer Service Representative for ordering information.
Specifications
Chemical Identifiers
Blown-vinyl Sponge
Specifications
Blown-vinyl Sponge
Gray
5 x 3 ft.
Product Suggestions
Product Suggestions