Learn More
Wearwell™ Tile-Top Antimicrobial Mats
Permanent antimicrobial surface destroys microorganisms on contact
Supplier: Wearwell™ 420.12X3X10AMBL
Description
Permanent antimicrobial surface destroys microorganisms on contact
- Ideal for use in all dry work areas from packaging, banks, labs, office and behind or in front of counters
- Unique antimicrobial Nitricell™ sponge base offers long life and resilience
- 0.5 in. (1.3cm) sponge base for extra comfort
- Extra wear layer for abrasion and chemical resistance
Chemical Identifiers
| 0.5 in. |
Specifications
| 3 x 10 ft. | |
| 0.5 in. | |
| Blue |