Learn More
Wearwell™ Tile-Top Antimicrobial Mats
Permanent antimicrobial surface destroys microorganisms on contact
Supplier:  Wearwell™ 420.12X2X3AMCH
 View more versions of this product
Catalog No. 191511211
Only null left
Description
Description
Permanent antimicrobial surface destroys microorganisms on contact
  • Ideal for use in all dry work areas from packaging, banks, labs, office and behind or in front of counters
  • Unique antimicrobial Nitricell™ sponge base offers long life and resilience
  • 0.5 in. (1.3cm) sponge base for extra comfort
  • Extra wear layer for abrasion and chemical resistance
Specifications
Chemical Identifiers
0.5 in.
Specifications
3 x 2 ft.
Charcoal Gray
0.5 in.
Product Suggestions
Product Suggestions