Learn More
Wearwell™ Rejuvenator™ Connect Antifatigue Mats and Accessories
100% urethane antifatigue mats, provide the ultimate long-term value
Supplier:  Wearwell™ 502.58X2X39MBK
 View more versions of this product
Catalog No. 191511222
Only null left
Description
Description
  • A perfect combination of extreme comfort, performance and exceptional resilience
  • Mat is literally crush resistant
  • Only modular urethane antifatigue mat
  • Mats connect on all sides to create an exceptional ergonomic floor
  • Optional 2 in. wide (5cm) yellow bevels available for increased safety (sold separately)
  • Dense exterior skin is tear and abrasion resistant
  • Thickness: 0.625 in. (1.6cm)
Warranty
10 year resilience
Specifications
Chemical Identifiers
Solid Anti-fatigue Floor Mat
Dry environments in Industrial, Lab and Pharma
Specifications
24 x 468 in.
Male ramp
Black
Solid Anti-fatigue Floor Mat
Dry environments in Industrial, Lab and Pharma
Product Suggestions
Product Suggestions