Learn More
Wearwell™ Grade A No. 482 Food Production Mats
Catalog No. p-3081253
Change view
Click to view available options
Dimensions (L x W):
30 x 36 in.
30 x 60 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
191500075 30 x 36 in.
191500076 30 x 60 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 191500075 Supplier Wearwell™ Supplier No. 482.12X30X36TC
Only null left
First antifatigue mat designed specifically for food production plants
  • Specially formulated nitrile rubber composite is 100% anti-microbial and withstands animal fats, fish and vegetable oils, and compounds used in a wide variety of food processing
  • Built-in handles ease cleaning of mats
  • Hygenic porthole and solid underside design simplify cleaning and virtually eliminate areas where food and bacteria can collect
  • Substantial underside knobs for increased comfort
  • Textured surface increases traction when greasy and wet
  • Terra cotta
Compliance
Meet NSF antimicrobial test ASTM™ G21-96 (2002).
Specifications
Dimensions (L x W) 30 x 36 in.