missing translation for 'onlineSavingsMsg'
Learn More

Wearwell™ Edgings for ErgoDeck™ Open Grid Flooring

$297.51

Specifications

Color Yellow
For Use With (Application) For wet, non-greasy areas
View More Specs

Products 1
Catalog Number Mfr. No. Style Price Quantity & Availability  
Catalog Number Mfr. No. Style Price Quantity & Availability  
19060515
View Documents Promotion Details
Wearwell™
560.78X6X18YL CS10
Side Ramps
Case of 10 Each for $297.51
Only null left
 
Description

Description

Specifications

Specifications

Yellow
For wet, non-greasy areas
Videos
SDS
Documents

Documents