Learn More
Wearwell™ Diamond-Plate SponeCote Work Mat with Gritshield Surface
Combines productivity improvement with slip prevention in oily overspray applications
$200.26 - $465.75
Description
- Nitrile enhanced Nitricell sponge base absorbs repeated compression
- Available in Diamond-Plate with GRITSHIELD UltraSoft and GRITSHIELD surface
- Machine cut and beveled edges prevent tripping
- Maximized standing area
- Overall Thickness: 15/16 in. (24mm)