Learn More
Watson-Marlow Marprene Double Segment Manifold Tubing
Supplier:  Watson-Marlow 979 0025 000
Catalog No. 14283168
Only null left
Description
Description
  • Made from autoclavable marprene
  • For use with 400DM2 and 400DM3 pumpheads
  • Can be used with any multichannel cartridges
  • Adjacent cartridges may contain tubing of a different material or size
Specifications
Specifications
0.25 mm
400DM2 and 400DM3 pumpheads
Manifold Tubing
Marprene
0.25 mm
Product Suggestions
Product Suggestions