Learn More
Waters Limited Spherisorb C6 Guard Cartridge, 80, 5 m, 4.6 mm X 10 mm, 3/pk
Catalog No. 50784286
Encompass
Spherisorb C6 columns are general purpose, silica-based reversed-phase columns that contain a linear-hexyl ligand. These USP L15 columns are used when you require a less retentive sorbent compared to the Spheriorb ODS columns.
Catalog No. 50784286 Supplier Waters Limited Supplier No. PSS830075
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Spherisorb C6 columns are general purpose, silica-based reversed-phase columns that contain a linear-hexyl ligand. These USP L15 columns are used when you require a less retentive sorbent compared to the Spheriorb ODS columns.