missing translation for 'onlineSavingsMsg'
Learn More

Water for TOC analysis, MilliporeSigma™ Supelco™

Catalog No. 111015497
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015497 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015497 Supplier Merck Emd Millipore Supplier No. 785331L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Total organic carbon analysis is a methodology to measure the carbon content of dissolved and particulate organic matter present in water. TOC analysis helps removal of inorganic carbon, oxidation of organic carbon into CO2 and quantifying CO2.

Environmental Food and Beverages and General Analytical

Chemical Identifiers

MDL Number MFCD00011332

Specifications

CAS 7732-18-5
Density 1.000 g/mL (at 3.98°C)
Boiling Point 100°C
Linear Formula H2O
Quantity 1 L
Vapor Density <1 (vs. air)
Vapor Pressure 3 mmHg
Formula Weight 18.02
ShelfLife Limited shelf life, expiry date on the label
RTECSNumber ZC0110000