missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water Solution, Contains 10 mM Ammonium Acetate, For HPLC, MilliporeSigma™ Supelco™
Specifications
Specifications
| Density | 0.996 g/mL (at 25°C) |
| Packaging | Glass Bottle |
| Linear Formula | H2O |
| Quantity | 4 x 4 L |
| MDL Number | MFCD00011332 |
| Grade | For HPLC |