missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water Solution, Contains 0.1% (v/v) Trifluoroacetic Acid, For HPLC, MilliporeSigma™ Supelco™
Description
The product is water containing 0.1% v/v trifluoroacetic acid. This pre-blended mobile phase solvent is of high purity that reduces time-consuming preparation and prevents instrument downtime because of impure mobile phases. It can be employed for HPLC and UHPLC (Ultra High performance Liquid Chromatography) techniques. Trifluoroacetic acid (TFA) is commonly used as a mobile phase additive. It is a UV transparent, volatile acid that forms hydrophobic negatively charged trifluoroacetate ion.
Specifications
Specifications
| Packaging | Nowpak™ |
| Linear Formula | H2O |
| MDL Number | MFCD00011332 |
| Quantity | 4 x 4 L |
| Grade | For HPLC |