missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Water Resistant Mid-Arm Length Cryogenic Gloves

Catalog No. 17355049
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17355049 Small
17355050 Medium
17355051 Large
17355052 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17355049 Supplier National Safety Apparel™ Supplier No. G99CRBERSMMA

Gloves provide protection from ultra-cold environments in research labs, frozen food processing, dry ice blasting, and more.

  • Gloves offer protection from ultra-cold environments in a wide variety of applications
  • Teflon™ Laminated Nylon with 3M™ Scotchlite™ Thinsulate™ liner
  • Water resistant, breathable membrane for additional splash protection
  • Knit wrist cuffs
  • Made in the USA

Specifications

Certifications/Compliance (Europe) EN 511 Contact Cold Performance Level 2, EN 511 Convective Cold Performance Level 3
Chlorinated No
Description Mid-Arm Length
Color Blue
Exterior Finish Smooth
Hand Specific
Material Nylon
Product Type Cryogenic Gloves
Size Small
For Use With (Application) Cryogenic Liquids/Gases
Glove Cuff Style Slip-on
Liner Material 3M™ Thinsulate™
Lining Lined
Series G99CRBER_ _MA
Length (English) 15 in.
Length (Metric) 38 cm
Product Line Water Resistant Cryogenic Gloves
Show More Show Less