missing translation for 'onlineSavingsMsg'
Learn More

Water for Injection, USP Grade*, G-Biosciences

Catalog No. S001119
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
1 gal.
10 L
50 L
100 L
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S001119 100 mL
S001102 1 L
S001104 50 L
S001126 10 L
S001120 500 mL
S001127 100 L
S001124 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. S001119 Supplier G-Biosciences Supplier No. 7862227

Water for Injection (WFI) USP Grade*, is produced in ISO9001:2015 and ISO 13485: 2016 certified facilities.

Meets specifications of ASTM (American Society for Testing and Materials) Type I Ultrapure water which is used for the most critical applications and advanced analytical procedures in the laboratory including cell and tissue culture, gas and liquid chromatography. Water for Injection (WFI) USP Grade* is tested for Conductivity, Sterility, Total Organic Carbon (TOC) and Endotoxin to meet the following specifications.

  • Conductivity: <1.3 μS/cm at 25°C
  • Sterility (Bacterial count): <10 cfu/100 mL
  • Total Organic Carbon (TOC): <500 ppb
  • Endotoxin: <0.25 IU/mL

* Endotoxin testing is done with the modified Limulus Amebocyte Lysate (LAL) Test. Sterility test is done with USP certified Tryptic Soy Broth (TSB) media with incubation at 37°C for 14 days.

Chemical Identifiers

Molecular Weight (g/mol) 18.02

Specifications

Percent Purity >99%
Grade USP
Sterility Sterile
Quantity 100 mL
Chemical Name or Material H2O