missing translation for 'onlineSavingsMsg'
Learn More

Water Hardness Indicator 1% Eriochrome Black T in 20% Triethanolamine/80% Isopropanol, Reagents

Catalog No. 01333383
Change view
Click to view available options
Quantity:
120 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333383 120 mL
01333382 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01333383 Supplier Labchem Inc Supplier No. CW200600120C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Water Hardness Indicator 1% Eriochrome Black T in 20% Triethanolamine/80% Isopropanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Color Dark Red
Volume (Metric) 120 mL
For Use With (Application) Water Analysis
Packaging Glass Bottle
Quantity 120 mL
Sterility Non-sterile