missing translation for 'onlineSavingsMsg'
Learn More

Water Hardness Buffer Without Magnesium EDTA, Reagents

Water Hardness Buffer Without Magnesium EDTA, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$157.11

Specifications

Color Colorless
For Use With (Application) Water Analysis
Packaging Poly Bottle
Sterility Non-sterile
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01333391
View Documents Promotion Details
N/A 1 L
Each for $157.11
Only null left
 
Description

Description

Specifications

Specifications

Colorless
Water Analysis
Poly Bottle
Non-sterile
Videos
SDS
Documents

Documents