missing translation for 'onlineSavingsMsg'
Learn More

Water Hardness Buffer Without Magnesium EDTA, Reagents

Catalog No. 01333391
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333391 1 L
01333390 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01333391 Supplier Labchem Inc Supplier No. CW0007001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Water Hardness Buffer Without Magnesium EDTA, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Color Colorless
Volume (Metric) 1 L
For Use With (Application) Water Analysis
Packaging Poly Bottle
Quantity 1 L
Sterility Non-sterile