missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water Hardness Buffer, Odorless, Reagents
Water Hardness Buffer, Odorless, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Specifications
| Color | Colorless to Slight Yellow |
|---|---|
| For Use With (Application) | Water Analysis |
| Packaging | Poly Bottle |
| Sterility | Non-sterile |