missing translation for 'onlineSavingsMsg'
Learn More

Water Hardness Buffer with Magnesium EDTA, Reagents

Water Hardness Buffer with Magnesium EDTA, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$130.54 - $197.71

Specifications

Color Colorless
For Use With (Application) Water Analysis
Sterility Non-sterile
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01333395
View Documents Promotion Details
N/A 1 L
Each for $197.71
Only null left
 
01333392
View Documents Promotion Details
N/A 500 mL
Each for $130.54
Only null left
 
Description

Description

Specifications

Specifications

Colorless
Water Analysis
Non-sterile
Videos
SDS
Documents

Documents