missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water Hardness Buffer, with Magnesium EDTA, Ricca Chemical
APHA/ASTM/EPA for water hardness testing.
Supplier: Ricca Chemical Company 920016
Description
- For water hardness testing.
Chemical Identifiers
| 7 to 14 | |
| Non-sterile | |
| Water Hardness Buffer |
Specifications
| 7 to 14 | |
| Natural Poly Bottle | |
| Non-sterile | |
| Water Hardness Buffer |
| 500 mL | |
| 500 mL | |
| APHA/ASTM/EPA for Water Hardness | |
| 16.9 oz. |