missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Water For Injection (WFI) for Cell Culture
High quality water for use as a solvent in the preparation of cell culture media and laboratory reagents.
Supplier: Gibco™ A1287301
Description
- Gibco™ Water for Injection (WFI) for Cell Culture meets the USP monograph for Water for Injection packaged in bulk for commercial use elsewhere
- The water is prepared by distillation and filtration (0.1 μm), with no added substances
- In addition to meeting USP standards, each lot is quality tested for sterility, endotoxin, conductivity, pH, osmolality, and oxidizable substances
- Conductivity, total organic carbon, endotoxin and bioburden are tested daily; metals are tested monthly
- It can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions, and diluting buffers
- Should be stored at 15° to 30°C
Specifications
| 4 to 6 | |
| Sterile-filtered | |
| 500 mL | |
| Room Temperature |
| Cell Culture | |
| None | |
| Distilled and filtered (0.1 μm) | |
| Gibco |
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.