missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Water For Injection (WFI) for Cell Culture
GREENER_CHOICE

Catalog No. A1287304
Change view
Click to view available options
Quantity:
500 mL
1000 mL
10 x 500 mL
6 x 1000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1287304 6 x 1000 mL
A1287301 500 mL
A1287302 1000 mL
A1287303 10 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A1287304 Supplier Gibco™ Supplier No. A1287304

High quality water for use as a solvent in the preparation of cell culture media and laboratory reagents.

Gibco™ Water for Injection (WFI) for Cell Culture is high quality water for use as a solvent in the preparation of cell culture media and laboratory reagents. Available in many sizes to meet your needs, Gibco™ Water for Injection (WFI) for Cell Culture features:

  • Water For Injection Quality
  • Gibco™ quality and performance
  • Use in cell culture and molecular biology applications

Water for injection quality

Gibco™ Water for Injection (WFI) for Cell Culture meets the USP monograph for Water for Injection packaged in bulk for commercial use elsewhere. The water is prepared by distillation and filtration (0.1 μm), with no added substances. We monitor the water quality on a routine basis. Conductivity, total organic carbon, endotoxin and bioburden are tested daily; metals are tested monthly.

Gibco™ quality and performance

The performance and consistency expected from Gibco™ liquid media products depends on the high-quality water we use in manufacturing. This water is also bottled as Gibco™ Water for Injection (WFI) for Cell Culture. In addition to meeting USP standards, each lot is quality tested for sterility, endotoxin, conductivity, pH, osmolality, and oxidizable substances.

Use in cell culture and molecular biology applications

Gibco™ Water for Injection (WFI) for Cell Culture can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions and diluting buffers.

Product Use

For Research Use Only or Further Manufacturing. Not for use in diagnostic procedures.

cGMP Manufacturing and Quality System

Gibco™ Water for Injection (WFI) for Cell Culture is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Specifications

Chemical Name or Material Dihydrogen monoxide
pH 4 to 6
CAS 7732-18-5
Molecular Weight (g/mol) 18.02 g/mol
Grade Cell Culture
Packaging Bottle
Sterility Sterile-filtered
Treatment(s) None
Quantity 6 x 1000 mL
Purification Method Distilled and filtered (0.1 μm)
Shipping Condition Room Temperature
Product Line Gibco
Show More Show Less
What is the difference between Water for Injection (WFI) for Cell Culture, Cat. No. A12873XX and Distilled Water, Cat. No. 15230XXX?

These two products are the same water. However, Water for Injection (WFI) for Cell Culture has undergone more testing, including conductivity measurement as well as testing for nitrate and total organic carbon.

How is Water for Injection (WFI) for Cell Culture prepared?

Water for Injection (WFI) for Cell Culture is prepared by distillation and filtration (0.1 µm), with no added substances. It is labeled “For Research Use Only of Futher Manufacturing. Not for use in diagnostic procedures”.

For Research Use or Further Manufacturing. Not for use in diagnostic procedures.