missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Water For Injection (WFI) for Cell Culture
GREENER_CHOICE

Catalog No. A1287302
Encompass
Change view
Click to view available options
Quantity:
500 mL
1000 mL
10 x 500 mL
6 x 1000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1287302 1000 mL
A1287301 500 mL
A1287303 10 x 500 mL
A1287304 6 x 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A1287302 Supplier Gibco™ Supplier No. A1287302
Only null left

High quality water for use as a solvent in the preparation of cell culture media and laboratory reagents.

  • Gibco™ Water for Injection (WFI) for Cell Culture meets the USP monograph for Water for Injection packaged in bulk for commercial use elsewhere
  • The water is prepared by distillation and filtration (0.1 μm), with no added substances
  • In addition to meeting USP standards, each lot is quality tested for sterility, endotoxin, conductivity, pH, osmolality, and oxidizable substances
  • Conductivity, total organic carbon, endotoxin and bioburden are tested daily; metals are tested monthly
  • It can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions, and diluting buffers
  • Should be stored at 15° to 30°C

Specifications

pH 4 to 6
Grade Cell Culture
Packaging Bottle
Sterility Sterile-filtered
Treatment(s) None
Quantity 1000 mL
Purification Method Distilled and filtered (0.1 μm)
Shipping Condition Room Temperature
Product Line Gibco

For Research Use or Further Manufacturing. Not for use in diagnostic procedures.